Anti-KIR2DL3 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128831EA 750 CHF
USBI128831
Anti-KIR2DL3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KIR2DL3 (killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR