Anti-Kinesin 2C Mouse Monoclonal Antibody [clone: 1G2]

Lieferant: United States Biological
USBI128829EA 733 CHF
USBI128829
Anti-Kinesin 2C Mouse Monoclonal Antibody [clone: 1G2]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: Kinesin 2C
Clonality: monoclonal
Clone: 1G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR