Anti-KIFC1 Mouse Monoclonal Antibody [clone: 2B9]

Supplier: US Biological
USBI128825EA 702 CHF
USBI128825
Anti-KIFC1 Mouse Monoclonal Antibody [clone: 2B9]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: KIFC1 (kinesin family member C1)
Clonality: monoclonal
Clone: 2B9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR