Anti-KIF6 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128824EA 750 CHF
USBI128824
Anti-KIF6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KIF6 (kinesin family member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR