Anti-KIF26A Mouse Monoclonal Antibody [clone: 3C10]

Supplier: US Biological
USBI128820EA 702 CHF
USBI128820
Anti-KIF26A Mouse Monoclonal Antibody [clone: 3C10]
Antibodies
Type: Primary
Antigen: KIF26A (kinesin family member 26A)
Clonality: monoclonal
Clone: 3C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR