Anti-KIF26A Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI128819EA 788 CHF
USBI128819
Anti-KIF26A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KIF26A (kinesin family member 26A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR