Anti-KIF20A Mouse Polyclonal Antibody

Supplier: US Biological
USBI128815EA 702 CHF
USBI128815
Anti-KIF20A Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KIF20A (kinesin family member 20A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR