Anti-KIF1C Mouse Monoclonal Antibody [clone: 1F12]

Supplier: US Biological
USBI128814EA 702 CHF
USBI128814
Anti-KIF1C Mouse Monoclonal Antibody [clone: 1F12]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: KIF1C (kinesin family member 1C)
Clonality: monoclonal
Clone: 1F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR