Anti-KIF16B Mouse Monoclonal Antibody [clone: 2B5]

Supplier: US Biological
USBI128812EA 702 CHF
USBI128812
Anti-KIF16B Mouse Monoclonal Antibody [clone: 2B5]
Antibodies
Type: Primary
Antigen: KIF16B (kinesin family member 16B)
Clonality: monoclonal
Clone: 2B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR