Anti-KIF15 Mouse Monoclonal Antibody [clone: 4D4]

Lieferant: United States Biological
USBI128810EA 733 CHF
USBI128810
Anti-KIF15 Mouse Monoclonal Antibody [clone: 4D4]
Antikörper
Type: Primary
Antigen: KIF15 (kinesin family member 15)
Clonality: monoclonal
Clone: 4D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR