Anti-PEAK1 Mouse Monoclonal Antibody [clone: 3E7]

Lieferant: United States Biological
USBI128809EA 733 CHF
USBI128809
Anti-PEAK1 Mouse Monoclonal Antibody [clone: 3E7]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: PEAK1 (pseudopodium-enriched atypical kinase 1)
Clonality: monoclonal
Clone: 30000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR