Anti-KIAA1467 Mouse Polyclonal Antibody

Supplier: US Biological
USBI128799EA 702 CHF
USBI128799
Anti-KIAA1467 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KIAA1467 (KIAA1467)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR