Anti-KIAA1456 Mouse Polyclonal Antibody

Supplier: US Biological
USBI128798EA 702 CHF
USBI128798
Anti-KIAA1456 Mouse Polyclonal Antibody
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: KIAA1456 (KIAA1456)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR