Anti-CEMIP Mouse Monoclonal Antibody [clone: 3C12]

Supplier: US Biological
USBI128797EA 702 CHF
USBI128797
Anti-CEMIP Mouse Monoclonal Antibody [clone: 3C12]
Antibodies
Type: Primary
Antigen: CEMIP (cell migration inducing protein, hyaluronan binding)
Clonality: monoclonal
Clone: 3C12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR