Anti-FASTKD2 Mouse Monoclonal Antibody [clone: 1B11]

Supplier: US Biological
USBI128793EA 702 CHF
USBI128793
Anti-FASTKD2 Mouse Monoclonal Antibody [clone: 1B11]
Antibodies
Type: Primary
Antigen: FASTKD2 (FAST kinase domains 2)
Clonality: monoclonal
Clone: 1B11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR