Anti-CEP104 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI128792EA 733 CHF
USBI128792
Anti-CEP104 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: CEP104 (centrosomal protein 104kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR