Anti-WAPAL Mouse Monoclonal Antibody [clone: 2B3]

Supplier: US Biological
USBI128789EA 702 CHF
USBI128789
Anti-WAPAL Mouse Monoclonal Antibody [clone: 2B3]
Antibodies
Type: Primary
Antigen: WAPAL (wings apart-like homolog (Drosophila))
Clonality: monoclonal
Clone: 2B3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR