Anti-MKI67 Mouse Monoclonal Antibody [clone: 7B8]

Lieferant: United States Biological
USBI128783EA 733 CHF
USBI128783
Anti-MKI67 Mouse Monoclonal Antibody [clone: 7B8]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: monoclonal
Clone: 7B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR