Anti-KHSRP Mouse Monoclonal Antibody [clone: 4C10]

Supplier: US Biological
USBI128782EA 702 CHF
USBI128782
Anti-KHSRP Mouse Monoclonal Antibody [clone: 4C10]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: KHSRP (KH-type splicing regulatory protein)
Clonality: monoclonal
Clone: 4C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR