Anti-KHK Mouse Monoclonal Antibody [clone: 1H6-2B6]

Supplier: US Biological
USBI128781EA 702 CHF
USBI128781
Anti-KHK Mouse Monoclonal Antibody [clone: 1H6-2B6]
Antibodies
Recommended dilution: Immunochemistry: 1-10 ?g/ml

Type: Primary
Antigen: KHK (ketohexokinase (fructokinase))
Clonality: monoclonal
Clone: 1H6-2B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR