Drucken

Anti-KHDRBS1 Mouse Polyclonal Antibody

Lieferant: US Biological
USBI128776EA 702 CHF
USBI128776
Anti-KHDRBS1 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: KHDRBS1 (KH domain containing, RNA binding, signal transduction associated 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR