Anti-KRT33B Mouse Monoclonal Antibody [clone: 4A6-A7]

Supplier: US Biological
USBI128773EA 702 CHF
USBI128773
Anti-KRT33B Mouse Monoclonal Antibody [clone: 4A6-A7]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: KRT33B (keratin 33B)
Clonality: monoclonal
Clone: 4A6-A7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR