Anti-KRT17 Mouse Monoclonal Antibody [clone: 2D4-1G9]

Supplier: US Biological
USBI128772EA 702 CHF
USBI128772
Anti-KRT17 Mouse Monoclonal Antibody [clone: 2D4-1G9]
Antibodies
Recommended dilution: Immunofluorescence: 25 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: KRT17 (keratin 17)
Clonality: monoclonal
Clone: 2D4-1G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR