Anti-KEL Mouse Monoclonal Antibody [clone: 4B10]

Supplier: US Biological
USBI128771EA 702 CHF
USBI128771
Anti-KEL Mouse Monoclonal Antibody [clone: 4B10]
Antibodies
Type: Primary
Antigen: KEL (Kell blood group, metallo-endopeptidase)
Clonality: monoclonal
Clone: 4B10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR