Anti-KDM2B Mouse Monoclonal Antibody [clone: 5G1]

Supplier: US Biological
USBI128769EA 702 CHF
USBI128769
Anti-KDM2B Mouse Monoclonal Antibody [clone: 5G1]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: KDM2B (lysine (K)-specific demethylase 2B)
Clonality: monoclonal
Clone: 5G1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR