Anti-KCTD17 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI128760EA 733 CHF
USBI128760
Anti-KCTD17 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: KCTD17 (potassium channel tetramerization domain containing 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR