Anti-KCNQ5 Mouse Monoclonal Antibody [clone: 2E2]

Supplier: US Biological
USBI128757EA 702 CHF
USBI128757
Anti-KCNQ5 Mouse Monoclonal Antibody [clone: 2E2]
Antibodies
Type: Primary
Antigen: KCNQ5 (potassium voltage-gated channel, KQT-like subfamily, member 5)
Clonality: monoclonal
Clone: 200
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR