Anti-KCNQ4 Mouse Monoclonal Antibody [clone: 2H6]

Supplier: US Biological
USBI128756EA 702 CHF
USBI128756
Anti-KCNQ4 Mouse Monoclonal Antibody [clone: 2H6]
Antibodies
Type: Primary
Antigen: KCNQ4 (potassium voltage-gated channel, KQT-like subfamily, member 4)
Clonality: monoclonal
Clone: 2H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR