Anti-KCNMB4 Mouse Monoclonal Antibody [clone: 1G7]

Lieferant: United States Biological
USBI128755EA 733 CHF
USBI128755
Anti-KCNMB4 Mouse Monoclonal Antibody [clone: 1G7]
Antikörper
Type: Primary
Antigen: KCNMB4 (potassium large conductance calcium-activated channel, subfamily M, beta member 4)
Clonality: monoclonal
Clone: 1G7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR