Anti-KCNMB3 Mouse Monoclonal Antibody [clone: 5H1]

Supplier: US Biological
USBI128752EA 702 CHF
USBI128752
Anti-KCNMB3 Mouse Monoclonal Antibody [clone: 5H1]
Antibodies
Type: Primary
Antigen: KCNMB3 (potassium large conductance calcium-activated channel, subfamily M beta member 3)
Clonality: monoclonal
Clone: 5H1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR