Anti-KCNK5 Mouse Monoclonal Antibody [clone: 2B4]

Supplier: US Biological
USBI128751EA 702 CHF
USBI128751
Anti-KCNK5 Mouse Monoclonal Antibody [clone: 2B4]
Antibodies
Type: Primary
Antigen: KCNK5 (potassium channel, subfamily K, member 5)
Clonality: monoclonal
Clone: 2B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR