Drucken

Anti-KCNK10 Mouse Monoclonal Antibody [clone: 1C1]

Lieferant: US Biological
USBI128750EA 702 CHF
USBI128750
Anti-KCNK10 Mouse Monoclonal Antibody [clone: 1C1]
Antikörper
Type: Primary
Antigen: KCNK10 (potassium channel, subfamily K, member 10)
Clonality: monoclonal
Clone: 1C1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR