Anti-KCNJ10 Mouse Monoclonal Antibody [clone: 1C11]

Lieferant: United States Biological
USBI128747EA 733 CHF
USBI128747
Anti-KCNJ10 Mouse Monoclonal Antibody [clone: 1C11]
Antikörper
Type: Primary
Antigen: KCNJ10 (potassium inwardly-rectifying channel, subfamily J, member 10)
Clonality: monoclonal
Clone: 1C11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR