Anti-KCNIP4 Mouse Monoclonal Antibody [clone: 1A11]
Supplier: US Biological
USBI128746EA 702 CHF
USBI128746
Anti-KCNIP4 Mouse Monoclonal Antibody [clone: 1A11]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: KCNIP4 (Kv channel interacting protein 4)
Clonality: monoclonal
Clone: 1A11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human