Anti-KCNIP1 Mouse Monoclonal Antibody [clone: 3D9]

Supplier: US Biological
USBI128742EA 702 CHF
USBI128742
Anti-KCNIP1 Mouse Monoclonal Antibody [clone: 3D9]
Antibodies
Type: Primary
Antigen: KCNIP1 (Kv channel interacting protein 1)
Clonality: monoclonal
Clone: 3D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR