Anti-KCNH6 Mouse Monoclonal Antibody [clone: 3D9]

Supplier: US Biological
USBI128741EA 702 CHF
USBI128741
Anti-KCNH6 Mouse Monoclonal Antibody [clone: 3D9]
Antibodies
Type: Primary
Antigen: KCNH6 (potassium voltage-gated channel, subfamily H (eag-related), member 6)
Clonality: monoclonal
Clone: 3D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR