Anti-KCNG3 Mouse Monoclonal Antibody [clone: 5H2]

Supplier: US Biological
USBI128740EA 702 CHF
USBI128740
Anti-KCNG3 Mouse Monoclonal Antibody [clone: 5H2]
Antibodies
Type: Primary
Antigen: KCNG3 (potassium voltage-gated channel, subfamily G, member 3)
Clonality: monoclonal
Clone: 5H2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR