Anti-KCNE1 Mouse Monoclonal Antibody [clone: 5B12]

Supplier: US Biological
USBI128737EA 702 CHF
USBI128737
Anti-KCNE1 Mouse Monoclonal Antibody [clone: 5B12]
Antibodies
Type: Primary
Antigen: KCNE1 (potassium voltage-gated channel, Isk-related family, member 1)
Clonality: monoclonal
Clone: 5B12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR