Anti-KCNC3 Mouse Monoclonal Antibody [clone: 1C1]

Supplier: US Biological
USBI128735EA 702 CHF
USBI128735
Anti-KCNC3 Mouse Monoclonal Antibody [clone: 1C1]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: KCNC3 (potassium voltage-gated channel, Shaw-related subfamily, member 3)
Clonality: monoclonal
Clone: 1C1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR