Anti-KCNA3 Mouse Monoclonal Antibody [clone: 1D8]

Supplier: US Biological
USBI128734EA 702 CHF
USBI128734
Anti-KCNA3 Mouse Monoclonal Antibody [clone: 1D8]
Antibodies
Type: Primary
Antigen: KCNA3 (potassium voltage-gated channel, shaker-related subfamily, member 3)
Clonality: monoclonal
Clone: 1D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR