Anti-KCNA1 Mouse Monoclonal Antibody [clone: 2D8]

Supplier: United States Biological
USBI128733EA 659 CHF
USBI128733
Anti-KCNA1 Mouse Monoclonal Antibody [clone: 2D8]
Antibodies
Type: Primary
Antigen: KCNA1 (potassium voltage-gated channel, shaker-related subfamily, member 1 (episodic ataxia with myokymia))
Clonality: monoclonal
Clone: 2D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR