Anti-KCNA1 Mouse Polyclonal Antibody

Supplier: US Biological
USBI128732EA 702 CHF
USBI128732
Anti-KCNA1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KCNA1 (potassium voltage-gated channel, shaker-related subfamily, member 1 (episodic ataxia with myokymia))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR