Anti-KLHL41 Mouse Monoclonal Antibody [clone: 1H3]

Supplier: US Biological
USBI128729EA 702 CHF
USBI128729
Anti-KLHL41 Mouse Monoclonal Antibody [clone: 1H3]
Antibodies
Type: Primary
Antigen: KLHL41 (kelch-like family member 41)
Clonality: monoclonal
Clone: 1H3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR