Anti-KATNBL1 Mouse Monoclonal Antibody [clone: 1E7]

Supplier: US Biological
USBI128726EA 702 CHF
USBI128726
Anti-KATNBL1 Mouse Monoclonal Antibody [clone: 1E7]
Antibodies
Type: Primary
Antigen: KATNBL1 (katanin p80 subunit B-like 1)
Clonality: monoclonal
Clone: 10000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR