Anti-KATNA1 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI128723EA 733 CHF
USBI128723
Anti-KATNA1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KATNA1 (katanin p60 (ATPase containing) subunit A 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR