Anti-Kappa Light Chain Mouse Monoclonal Antibody [clone: 1F6]

Supplier: US Biological
USBI128720EA 702 CHF
USBI128720
Anti-Kappa Light Chain Mouse Monoclonal Antibody [clone: 1F6]
Antibodies
Type: Primary
Antigen: Kappa Light Chain
Clonality: monoclonal
Clone: 1F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR