Anti-TRIM28 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI128715EA 733 CHF
USBI128715
Anti-TRIM28 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: TRIM28 (tripartite motif containing 28)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR