Anti-KLK5 Mouse Monoclonal Antibody [clone: 3H3]

Supplier: US Biological
USBI128714EA 702 CHF
USBI128714
Anti-KLK5 Mouse Monoclonal Antibody [clone: 3H3]
Antibodies
Type: Primary
Antigen: KLK5 (kallikrein-related peptidase 5)
Clonality: monoclonal
Clone: 3H3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR