Anti-KLK14 Mouse Monoclonal Antibody [clone: 2A7]

Supplier: US Biological
USBI128712EA 702 CHF
USBI128712
Anti-KLK14 Mouse Monoclonal Antibody [clone: 2A7]
Antibodies
Type: Primary
Antigen: KLK14 (kallikrein-related peptidase 14)
Clonality: monoclonal
Clone: 2A7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR