Anti-KLK13 Mouse Monoclonal Antibody [clone: 1G9]

Supplier: US Biological
USBI128711EA 702 CHF
USBI128711
Anti-KLK13 Mouse Monoclonal Antibody [clone: 1G9]
Antibodies
Type: Primary
Antigen: KLK13 (kallikrein-related peptidase 13)
Clonality: monoclonal
Clone: 1G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR